Would you like to log in again?
Blood Bank Saline Solutions, Azer Scientific
Supplier: Azer Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
ES124310L ES124420L ES12444G ES124410L ES124320L ES12444G
16005-092CS 81.84 USD InStock
16005-092 CA16005-092 16005-094 16005-090 16005-096 16005-088
Blood Bank Saline Solutions, Azer Scientific
Buffers Cell Culture Buffers
Blood bank saline is pH-controlled for a pH between 6.5 and 7.0
  • Meets NCCLS standard ASI/BB-1
  • Both standard and buffered saline remain stable without preservatives
  • Cubes feature a spigot for easy dispensing
Buffered saline is tightly controlled to a pH range of 7.0 to 7.2 and is specially designed to enhance weak agglutination reactions in blood bank procedures.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty