Would you like to log in again?
Tychem® Hoods, Bullard®
Supplier: Bullard
Tychem®
20TICH RT3 20TJN RT3T 20SICH 20TICN 20TICSN 20TJ RT1 20SIC 20SICN 20TICS
89235-851CS 906.32 USD InStock
89235-851 16006-048 16005-040 16005-038 16006-050 16006-042 16006-053 16006-054 16006-040 16006-300 16006-044 89093-555
Tychem® Hoods, Bullard®
Respirators Respiratory Systems
Tychem® hoods provide a variety of simple, effective, and economical options for respiratory protection.
  • Simple, effective, and economical options for respiratory protection
  • Covers entire head, face, and neck to provide splash and overspray protection
  • Neck cuff design maintains positive pressure inside the hood by inflating the inner collar to provide a snug, comfortable fit and continuous airflow
  • Wide-angle lens offers large field of vision while accommodating beards, prescription eyeglasses, or safety glasses
  • Lighter and more comfortable than tight-fitting full facepiece respirators
  • Durable enough to reuse, yet economical enough to discard after each use
  • Approved for NIOSH for use in Type-C applications
These hoods do not require fit testing or extensive recordkeeping. Tychem® QC hoods are poly-coated and are ideal for use with pharmaceutical manufacturing, painting, healthcare applications, and general industry. Tychem® SL hoods all feature taped and sealed seams and are SARANEX® film-coated for use in applications where chemical protection properties are critical. Approved for NIOSH for use in Type C applications.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty