Would you like to log in again?
Volumetric Flasks, Class A, Wide Mouth, Heavy Duty, Chemglass
Supplier: Chemglass
CG-1615-50 CG-1615-25 CG-1615-20 CG-1615-250 CG-1615-05 CG-1615-10 CG-1615-200 CG-1615-100 CG-1615-2L CG-1615-500 CG-1615-1L
16007-058EA 125.28 USD InStock
16007-058 16007-048 16007-060 16007-050 16007-062 16007-052 16007-064 16007-054 16007-066 16007-056 16007-046
Volumetric Flasks, Class A, Wide Mouth, Heavy Duty, Chemglass
Flasks Volumetric Flasks
Wide mouth design facilitates mixing, pipette access, sampling and filling
Large permanent white letters make the capacity clearly visible. Flasks are calibrated "To Contain" at Class A tolerances, per ASTM E288 specifications. Supplied complete with either a [ST] glass pennyhead or PTFE stopper.
Unique Flat Bottom Interior permits the use of a standard stir bar.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty