Would you like to log in again?
Nalgene® PassPort™ IP2 Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene® PassPort™
2099-0010 2099-0002 2099-0004 2099-0016 2099-0008 2099-0001 2099-0032
16057-166CS 309.12 USD InStock
16057-166 16057-168 16057-169 16057-170 16057-160 16057-162 16057-164
Nalgene® PassPort™ IP2 Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Excellent for small-scale inter-lab shipping of hazardous samples and chemicals, such as corrosives and cleaning compounds.
  • Leakproof
  • With polypropylene screw closures
The application of pressure or vacuum may result in product failure, damage to property, or personal injury.
Recommended for customers who are designing, assembling, and certifying their own combination packaging. Equipped with heavy-duty walls, bottles are designed to resist splitting and puncturing from shipping shocks; proven effective by industrial users. IP2 marking, material, volume, registration notch, and NALGENE name are molded into the bottle's bottom. Evaluated at 15 psi (103 kPa) per 49CFR 173.27 (c) (2) ICAO Technical Instructions Part 3-1; 1.1.6.1, and IATA Dangerous Goods Regulations Section 5.0.2.9.
Ordering information: For replacement caps for 16057-169 and -170, see 16219-122 series.
Caution: Do not use bottle under pressure or vacuum.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty