Would you like to log in again?
Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
342024-0030 342024-1000 342024-0250 342020-1000 342024-0060 342024-0125 342020-9500 342020-0250 342020-2000 342020-0030 342020-0060 342024-0500 342020-2000 342020-0060 342020-0500 342020-0125
21010-402CS 317.14 USD InStock
21010-402 16059-490 CATX16059-914 16057-310 16057-302 16057-314 16057-304 16057-306 16057-308 16159-999 16059-912 16059-914 16059-498 CATX16159-999 16059-494 16059-496
Nalgene® Square Media Bottles, PETG, Sterile, Thermo Scientific
Bottles Plastic Bottles
Nalgene® polyethylene terephthalate copolyester (PETG) square media bottles are an inexpensive alternative to glass media bottles.
  • Available with or without a natural HDPE screw cap closure
  • Bottles and closures are not autoclavable
  • Break-resistant, durable, and leakproof
Transparency and reduced permeability to CO₂/O₂ allow media storage for up to six months. Bottles and closures are radiation-sterilized and nonpyrogenic. Bottle 16159-999 (2 L size) features molded-in handgrips.
Ordering information: Packaged in shrink-wrapped trays. For additional screw cap/closure options, see 16060-026 series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty