Would you like to log in again?
Nalgene® Laboratory Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2003-0001 2003-9050 2003-0016 2003-0032 2003-9025 2003-9016 2003-0002 2003-0008 2003-0004
16057-980CS 88.13 USD InStock
16057-980 16058-123 16058-145 16058-112 16058-101 16058-021 16058-043 16058-065 16058-000
Nalgene® Laboratory Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Leakproof, translucent, flexible bottles, with excellent impact resistance and better visibility to contents than HDPE bottles.
  • Excellent chemical resistance to most acids, bases, and alcohols
  • LDPE is extremely low in trace metal content
  • Excellent material for trace metal analysis
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: For replacement closures for 16058-112 and 16058-145, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty