Would you like to log in again?
Nalgene® Large Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2202-0005 2202-0010 2202-0020
16058-203CS 355.2 USD InStock
16058-203 16058-167 16058-189
Nalgene® Large Bottles, Low-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Large bottles are designed for collecting, storing, or shipping anything from distilled water to corrosive chemicals and standards.
  • With polypropylene closures
  • Leakproof
A built-in loop on the shoulder (except on the 2 L size) is convenient for attaching an identification tag.
Ordering information: For replacement closures, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty