Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nalgene® Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific

Supplier: Thermo Scientific

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

Nalgene®
2205-0032 DS2205-0210 2205-0016 DS2205-0250 DS2205-0010 DS2205-0020
16058-350EA 93.41 USD InStock
16058-350 16058-250 16058-382 16058-305 16058-258 16058-252
Nalgene® Laboratory Bottles, Polycarbonate, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Excellent impact resistance and clarity.

  • 2 and 2.5 L sizes are ideal for use as culture vessels for batch, suspension, or roller culturing
  • 3.8 and 8 L sizes have shoulder loop for attaching identification tags
  • Not recommended for vacuum applications
  • With polypropylene screw closures
  • Leakproof, Autoclavable

Note: Before autoclaving, set closure on top of bottle without engaging threads.

Ordering information: For replacement closures, see 16219-122 series.

Order Now
 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty