Would you like to log in again?
BOTTLE PFA GRAD NARW-MTH3000ML
Supplier: Saint Gobain Life Sciences
Chemware Bottle, Teflon PFA Graduated, Narrow Mouth^ Length/Height (w/Closure): 269.9 mm. Neck I.D. (Plug Measure): 36.6 mm. Capacity to Brim: 3350 ml. Leakproof, unbreakable PFA bottles for storage of virtually all corrosive chemicals, including hyfrofluoric, nitric and perchloric acids.
ORDER
Expand All / Collapse All
Description VWR Catalog Number Supplier No Unit Price Quantity
BOTTLE PFA GRAD NARW-MTH3000ML 16059-421 D1069073 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty