Would you like to log in again?
Nalgene® Narrow-Mouth Packaging Bottles, Translucent Amber HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312084-0004 312084-0016
16059-508CS 263.16 USD InStock
16059-508 16059-504
Nalgene® Narrow-Mouth Packaging Bottles, Translucent Amber HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
These translucent amber high density polyethylene bottles are leakproof.
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
They include opaque amber polypropylene closures. These bottles have excellent chemical resistance and have a robust construction for packaging use, and are designed for storage of light sensitive liquids.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty