Would you like to log in again?
Nalgene® Packaging Bottles, HDPE, Narrow-Mouth, with Screw Caps, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
312089-0008 312085-0004 312085-0002 312085-0016 312089-0001 312089-0002 312085-0001 312089-0004 312089-0032 312089-0016 312085-0008 312085-0032
16059-516CS 442.56 USD InStock
16059-516 16059-518 16059-512 16059-556 16059-548 16059-514 16059-558 16059-520 16059-552 16059-522 16059-554 16059-550
Nalgene® Packaging Bottles, HDPE, Narrow-Mouth, with Screw Caps, Thermo Scientific
Bottles Plastic Bottles
These economical, break-resistant bottles are made from high-density polyethylene.
  • Available in natural or amber colors
  • Guaranteed leakproof
Natural sample bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Ordering information: Supplied with polypropylene screw caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty