Would you like to log in again?
Nalgene® Narrow-Mouth Packaging Bottles, PPCO with Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312087-0001 312087-0004 312087-0002 312087-0032 312087-0008 312087-0016
16059-528CS 512.64 USD InStock
16059-528 16059-524 16059-534 16059-526 16059-530 16059-532
Nalgene® Narrow-Mouth Packaging Bottles, PPCO with Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
These narrow mouth polypropylene copolymer bottles have a robust construction for packaging use and are featured with polypropylene closures.
  • Exhibit excellent impact and chemical resistance and clarity
  • Leak proof and autoclavable and have a translucent color
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty