Would you like to log in again?
Nalgene® LDPE Narrow Mouth Packaging Bottles, with PP Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312088-0032 312088-0004 312088-0016
16059-546CS 166.15 USD InStock
16059-546 16059-544 16059-540
Nalgene® LDPE Narrow Mouth Packaging Bottles, with PP Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene low density polyethylene narrow mouth packaging bottles have a robust construction for packaging use and are featured with polypropylene closures.
These packaging bottles have excellent impact chemical resistance and clarity. They are constructed of the ideal material for trace metal analysis
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty