Would you like to log in again?
Nalgene® Packaging Bottles, Amber, Wide Mouth, Translucent, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312184-0008 312184-0004 312184-0016
16059-568CS 288 USD InStock
16059-568 16059-564 16059-566
Nalgene® Packaging Bottles, Amber, Wide Mouth, Translucent, HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Amber wide-mouth packaging bottles are high density polyethylene.
The amber color protects light sensitive products, and the wide mouth allows for easy filling and dispensing of liquids and powders. The bottles come with opaque amber polypropylene closures.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty