Would you like to log in again?
Nalgene Wide-Mouth Packaging Bottles, PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312187-0008 312187-0032 312187-0004 312187-0001 312187-0016 312187-0002
16059-590CS 445.82 USD InStock
16059-590 16059-586 16059-588 16059-592 16059-584 16059-594
Nalgene Wide-Mouth Packaging Bottles, PPCO, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® wide-mouth polypropylene copolymer packaging bottles are leak proof and have wide-mouths which allows for easy filling and dispensing of powders.
These bottles come with polypropylene closures
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty