Would you like to log in again?
Nalgene® Packaging Bottles, Wide Mouth, HDPE, with Closure, Bulk Packed, Thermo Scientific
Supplier: Thermo Scientific
312185-0002 312185-0008 312185-0032 312185-0004 312189-0002 312185-0016 312189-0016 312189-0032 312185-0001 312189-0004 312189-0001 312189-0008
16059-604CS 286.55 USD InStock
16059-604 16059-580 16059-606 16059-578 16059-600 16059-602 16059-574 16059-596 16059-576 16059-598 16059-582 16059-572
Nalgene® Packaging Bottles, Wide Mouth, HDPE, with Closure, Bulk Packed, Thermo Scientific
Bottles Plastic Bottles
These economical, break-resistant bottles are made from high-density polyethylene.
  • Available in natural or amber colors
  • Guaranteed leakproof
Natural sample bottles allow easy viewing of liquid level. Amber bottles reduce UV light transmission for protection of light-sensitive liquids.
Ordering information: Supplied with polypropylene screw caps.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty