Would you like to log in again?
Nalgene® Diagnostic Bottles, with Closure, Natural PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312006-9125 312006-9025 312006-9050
16059-668CS 1518.72 USD InStock
16059-668 16059-650 16059-660
Nalgene® Diagnostic Bottles, with Closure, Natural PPCO, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Thermo Scientific Nalgene PPCO diagnostic bottles with polypropylene closures are used in test kits and many small-volume applications.
Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty