Would you like to log in again?
Nalgene® Diagnostic Bottles, Translucent Amber, HDPE, with Closure, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312084-9050 312084-9025 312084-9125
16059-662CS 1617.14 USD InStock
16059-662 16059-654 16059-670
Nalgene® Diagnostic Bottles, Translucent Amber, HDPE, with Closure, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene translucent amber high density polyethylene diagnostic bottles come with opaque amber polypropylene closures.
These bottles are used in test kits and many small-volume applications. Diagnostic bottles can be used for sample storage and shipping of small volume, valuable reagents that are light sensitive.
Ordering information: Bulk packed for packaging applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty