Would you like to log in again?
Nalgene® Diagnostic Bottles, HDPE, with Closures, Non-Sterile, Tray Pack, Thermo Scientific
Supplier: Thermo Scientific
322002-9025 322002-9125 322004-9125 322002-9025 322004-9025 322002-9050 322004-9050
16059-678CS 1172.16 USD InStock
16059-678 16059-688 16059-674 16059-696 CA10098-580 10098-580 16059-694
Nalgene® Diagnostic Bottles, HDPE, with Closures, Non-Sterile, Tray Pack, Thermo Scientific
Bottles Plastic Bottles
Perform many small volume applications with safe and leakproof Thermo Scientific™ Nalgene™ HDPE diagnostic bottles with closures.
  • Available in natural and amber
These bottles are ideal for use in test kits, sample storage and shipping of small volume, valuable reagents.
Certifications: Amber bottles meet the requirements of light-resistant containers per the current edition of USP.
Packaging: They come packed in rigid trays for easy handling.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty