Would you like to log in again?
Nalgene® IP2 High Density Polyethylene Wide Mouth Bottles, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312199-0032 312199-0008 312199-0016 312199-0004
16059-726CS 1412.16 USD InStock
16059-726 16059-728 16059-732 16059-730
Nalgene® IP2 High Density Polyethylene Wide Mouth Bottles, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® IP2 high density polyethylene wide-mouth bottles are leak proof and meet ICAO and IATA requirements.
These bottles are safe, economical and are for customers designing, assembling and certifying combination packaging. The bottles are recommended for storage and shipment of liquid reagents.
Ordering information: Bottles and closures are bulk packed in separate polybags within the same carton.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty