Would you like to log in again?
Nalgene® Square Media Bottles with Closures, PETG, Nonsterile Tray Pack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
322020-2000 322020-1000 322020-0030 322020-0060 322020-0250 322020-0125 322020-0500
16059-736CS 484.84 USD InStock
16059-736 16059-738 16059-744 16059-746 16059-734 16059-740 16059-742
Nalgene® Square Media Bottles with Closures, PETG, Nonsterile Tray Pack, Thermo Scientific
Bottles Cell Culture Bottles
Nalgene® polyethylene terephthalate copolyester square media bottles are specifically designed for packaging and shipping liquid media, buffers, and sera.
  • Non sterile
  • Leak proof
  • Molded-in graduations
These bottles are heavy-walled, durable, shatter-resistant, and provide excellent gas barrier properties.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty