Would you like to log in again?
Nalgene® Boston Round Bottles, HDPE, Narrow Mouth, Bulk Packed, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
312004-0008 312002-9050 312004-0016 312002-0004 312002-9125 312002-0002 312004-9050 312004-0001 312002-0008 312004-9125 312004-9025 312002-0032 312004-0002 312002-9025 312002-0006 312004-0032 312002-0016 312002-0001 312004-0004
16059-802CS 577.92 USD InStock
16059-802 16059-804 16059-766 16059-656 16059-666 16059-776 16059-798 16059-768 16059-658 16059-800 16059-646 16059-794 16059-772 16059-764 16059-664 16059-642 16059-796 16059-774 16059-770
Nalgene® Boston Round Bottles, HDPE, Narrow Mouth, Bulk Packed, Thermo Scientific
Bottles Plastic Bottles
These high-density polyethylene bottles are ideal for storing, packaging, and shipping liquids
  • Bottles are available in both translucent and amber formats
  • Translucence allows easy viewing of liquid levels
  • Amber reduces UV light transmissions to protect light-sensitive liquids
They offer excellent chemical resistance, and are highly reliable and durable for long-term use. Leakproof.
Certifications: Manufactured in compliance with ISO 13485:2003 quality requirements. Diagnostic Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Class VI.
Packaging: Bulk packed for packaging applications. Bottles and closures are packed in separate polybags within the same carton.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty