Would you like to log in again?
Nalgene® Narrow-Mouth Boston Round Bottles, Natural PPCO, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
312006-0016 312006-0002 312006-0008 312006-0004 312006-0032 312006-0001
16059-814CS 578.88 USD InStock
16059-814 16059-816 16059-810 16059-812 16059-806 16059-808
Nalgene® Narrow-Mouth Boston Round Bottles, Natural PPCO, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
These polypropylene copolymer narrow-mouth, Boston round bottles are feature polypropylene closures.
  • Durable
  • Translucent
  • Autoclavable
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty