Would you like to log in again?
Nalgene® Rectangular Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
312009-0032 312007-0004 312007-0032 312009-0004 312009-0008 312007-0016 312007-0008
16059-824CS 469.44 USD InStock
16059-824 16059-826 16059-832 16059-820 16059-822 16059-818 16059-828
Nalgene® Rectangular Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene® high density polyethylene rectangular bottles come with polypropylene closures.
Bottles are available in both translucent and amber. Translucence allows easy viewing of liquid levels. Amber reduces UV light transmissions to protect light-sensitive liquids.
All the bottles are leak proof, and feature a space-saving rectangular design. These bottles feature uniform walls which are resistant to splitting or puncturing.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Polypropylene screw closures are included.
Packaging: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty