Would you like to log in again?
Nalgene® Bottles, Wide Mouth, Natural LDPE, with PP Closure, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312103-0004 312103-0016
16059-858CS 1325.76 USD InStock
16059-858 16059-862
Nalgene® Bottles, Wide Mouth, Natural LDPE, with PP Closure, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene wide-mouth bottles are leakproof and constructed of natural low-density polyethylene.
These translucent bottles offer durability, clarity and chemical resistance. The bottles also contain uniform walls that resist splitting and puncturing.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty