Would you like to log in again?
Nalgene Wide-Mouth Bottles, Natural PPCO with Polypropylene Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312105-0001 312105-0016 312105-0008 312105-0004 312105-0032
16059-886CS 767.81 USD InStock
16059-886 16059-878 16059-888 16059-882 16059-884
Nalgene Wide-Mouth Bottles, Natural PPCO with Polypropylene Closures, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® Wide-Mouth polypropylene copolymer bottles are leak proof, auotclavable.
  • Feature a wide-mouth for ease of filling and dispensing
  • Bottles are translucent which offers better clarity than HDPE
These bottles have uniform walls which resist splitting and puncturing.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty