Would you like to log in again?
Nalgene® Wide Mouth Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312106-0001 312106-0016 312104-0032 312104-0004 312104-0002 312106-0004 312104-0001 312104-0016 312106-0032 312106-0008 312104-0008
16059-868CS 1774.08 USD InStock
16059-868 16059-898 16059-876 16059-900 16059-866 16059-894 16059-872 16059-874 16059-896 16059-890 16059-870
Nalgene® Wide Mouth Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® high-density polyethylene bottles have excellent chemical resistance, and are guaranteed leak proof.
  • Feature a wide-mouth for ease of filling and dispensing
  • Bottles have uniform walls that resist splitting and puncturing
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: Bottles are bulk packed. Available in natural and amber. Amber bottles comply with U.S. Pharmacopoeia 23,661 requirements for maximum allowable light transmission.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty