Would you like to log in again?
Nalgene® Wide-Mouth Square Bottles, HDPE, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
312114-0016 312114-0008 312114-0032 312114-0006 312114-0002
16059-902CS 1749.12 USD InStock
16059-902 16059-904 16059-910 16059-906 16059-908
Nalgene® Wide-Mouth Square Bottles, HDPE, Bulk Pack, Thermo Scientific
Bottles Roller Bottles
Nalgene® wide-mouth high density polyethylene square bottles are leak proof and have a square shape that saves shelf space.
The bottles have uniform walls that are resistant to splitting or puncturing, and have a wide mouth for easy filling and pouring.
Certifications: Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
Ordering information: They are bulk packed for packaging applications.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty