Would you like to log in again?
Nalgene® High-Density Polyethylene Economy Bottles, Bulk Pack, Wide Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
332189-0008 332189-0032 332189-0016 332189-0004 332189-0002
16060-008CS 691.2 USD InStock
16060-008 16060-010 16060-014 16060-012 73520-258
Nalgene® High-Density Polyethylene Economy Bottles, Bulk Pack, Wide Mouth, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Economy Bottles are lightweight, more flexible versions of the original Nalgene Lab Quality Bottles for less rigorous lab use.
  • Priced below low-quality plastic or glass bottle brands
  • Economical, quality choice for lighter-duty lab use
These Nalgene® Economy Bottles feature excellent impact and chemical resistance and clarity, include a polypropylene closure, and are made from natural, translucent high-density polyethylene. Like most Nalgene bottles, the Economy Bottles are guaranteed leakproof and are made in our ISO-certified USA manufacturing facility.
These bottles are suitable for light-duty, general purpose laboratory applications such as containing samples and daily working solutions like buffers and bench reagents.
Ordering information: Packaged in bulk quantities. Direct UV exposure should be avoided.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty