Would you like to log in again?
Nalgene® Diagnostic Bottles, Natural PPCO, without Closures, Bulk pack, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
362006-9125 362006-9050 362006-9025
16060-032CS 1001.28 USD InStock
16060-032 16060-030 16060-034
Nalgene® Diagnostic Bottles, Natural PPCO, without Closures, Bulk pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene natural PPCO diagnostic bottles are used in test kits and many small-volume applications.
The diagnostic bottles are non-sterile, and are made of polypropylene copolymer. They can be used for sample storage and shipping of small volume, valuable reagents. Closures sold separately.
Ordering information: Bulk Packed for Packaging Applications. For polypropylene caps, see 16060-026 series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty