Would you like to log in again?
Nalgene® Diagnostic Bottles, Opaque Amber, HDPE, without Closures, Bulk Pack, Thermo Scientific
Supplier: Thermo Scientific
362004-9125 362004-9050 362004-9025
16060-042CS 1086.7 USD InStock
16060-042 16060-040 16060-044
Nalgene® Diagnostic Bottles, Opaque Amber, HDPE, without Closures, Bulk Pack, Thermo Scientific
Bottles Plastic Bottles
Nalgene opaque amber high density polyethylene diagnostic bottles are used in test kits and many small-volume applications.
They have durable uniform walls that are resistant to splitting or puncturing. Diagnostic bottles can be used for sample storage, and shipping of valuable reagents that are light sensitive. Bulk pack comes without closures.
Ordering information: Bulk packed for packaging applications. For polypropylene caps, see 16060-026 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty