Would you like to log in again?
Nalgene® Large Polypropylene Copolymer Bottles, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2204-0020 2203-0010 2204-0010 2204-0005 2203-0020 2203-0005
16062-186CS 432.96 USD InStock
16062-186 16062-164 16062-142 16067-168 16067-146 16067-180
Nalgene® Large Polypropylene Copolymer Bottles, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Thermo Scientific Nalgene® Large Narrow Mouth Bottles feature excellent chemical resistance to most acids, bases, and alcohols, making them ideal for storing chemicals and standards as well as collecting and storing distilled water.
The 3.8 L and 8 L sizes feature a built-in shoulder loop for attaching an identification tag. All bottles are supplied with polypropylene screw closures. The 8 L bottle includes a white PP closure with a TPE gasket.
Avavilable in natural and amber finishes. Amber bottles comply with U.S. Pharmacopoeia 23, 661 requirements for maximum allowable light transmission.
They can be autoclaved repeatedly with or without contents.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty