Would you like to log in again?
Nalgene® Boston Round Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
Nalgene®
2006-9125 2006-0032 2006-0002 2006-9050 2006-9025 2006-0001 2006-0008 2006-0016 2006-0004
16066-960CS 114.55 USD InStock
16066-960 16067-124 16066-981 16067-102 16067-044 16067-000 16067-022 16067-088 16067-066
Nalgene® Boston Round Bottles, Polypropylene Copolymer, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Ideal for lab applications.
  • Excellent chemical resistance
  • With polypropylene screw closures
  • Leakproof, Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.
The 3.8 and 8 L sizes have a built-in shoulder loop for attaching an identification tag.
Ordering information: For replacement closures, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty