Would you like to log in again?
Nalgene® Boston Round Bottles, Teflon® Resin FEP, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
1600-0064 1600-0032 1600-0001 1600-0008 1600-0002 1600-0016 1600-0004
16071-020CS 623.04 USD InStock
16071-020 16071-085 16071-041 16071-008 16071-245 16071-165 16071-121
Nalgene® Boston Round Bottles, Teflon® Resin FEP, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
The most chemical and corrosion-resistant bottle available.
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set cap on top of bottle without engaging threads.
Excellent for high- and low-temperature work, trace metal analysis, and many applications with organic solvents. For high-purity analysis or storage, these bottles can be vigorously cleaned in boiling nitric acid. Bottle with Tefzel® ETFE screw cap withstands temperatures from –105 to 150 °C (–157 to 302 °F).
Packaging: Packaged individually.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty