Would you like to log in again?
Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
1630-0004 1630-0032 DS1630-0002 DS1630-0001 1630-0008 1630-0016
16071-570CS 1822.08 USD InStock
16071-570 16071-568 16071-566 16071-563 16071-564 16071-561
Nalgene® Laboratory Bottles, Teflon® PFA, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Teflon® PFA resists temperature, chemicals, and corrosion.
  • Withstand temperatures from –270 to 250 °C (–454 to 482 °F)
  • Translucent bottles allow liquid levels to be seen
  • Linerless, leakproof closures made of Teflon PFA for a positive seal
  • Leakproof, Autoclavable
Note: Before autoclaving, set closure on top of bottle without engaging threads.
Inert to virtually all chemicals except molten alkali metals, fluorine at elevated temperatures, and certain complex halogenated compounds at elevated temperatures and pressures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty