Would you like to log in again?
Modern Round Bottles, Nylon/PE, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLC-03741 PLC-03744
16085-160CS 71.26 USD InStock
16085-160 16085-181
Modern Round Bottles, Nylon/PE, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Nylon/PE barrier modern round bottles are nylon bottles co-extruded with PE & banded together.
This barrier prevents trace ingredients getting into the 'sidewall maze'. Containers produced by this process offer improved protection for non-polar hydrocarbons.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty