Would you like to log in again?
Lab Style Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-03155 PLA-03149 PLA-03153 PLA-03159 251953 501138 PLA-03127 249619 PLA-03157
16086-016CS 104.42 USD InStock
16086-016 16086-048 16086-028 16086-060 80080-922 16086-032 93001-664 76289-364 76003-538
Lab Style Bottles, High-Density Polyethylene, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Narrow mouth HDPE lab style bottles are supplied with linerless polypropylene caps.
Excellent chemical resistance and the leak resistant closure make these ideal for storing, shipping, and packaging liquids for long periods. HDPE bottles are ideal for high-speed filling, demanding pharmaceutical specifications and the rough handling of interstate shipping. Linerless white polypropylene caps offer excellent impact strength, cost effectiveness, and pliability. Caps attached to the convenience pack and enclosed on the bulk pack
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty