Would you like to log in again?
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Supplier: Qorpak
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
GLC-00999 GLC-05185 GLC-01006 GLC-01009 GLC-04878 GLC-07467 GLC-00985 GLC-05027 GLC-06064 GLC-00990 GLA-00788 GLC-04879 GLC-04835 GLC-04893 GLC-08765 GLC-01039 GLC-00991 GLC-01031 GLC-01016 GLC-05028 GLA-00789 GLC-07876 GLA-07795 GLC-07367 GLA-00794 GLA-00786 GLC-01022 GLA-00793 GLC-00986 GLC-04881 GLC-04995 GLC-04882 GLC-05183 GLA-00792 GLC-08764 242633 GLC-06994 GLC-01049 GLA-07790 GLC-00998 GLC-06160 GLC-00995 GLC-04925 GLC-09098 GLC-00982 GLC-07863 GLC-01050 GLC-01013 GLC-01014 GLC-00996 GLC-00981 GLC-09126 GLC-01001 GLC-04884 283154 GLA-00791 GLA-00790 GLC-04947 GLC-00987 GLC-01002 GLC-06849 GLC-00994 GLC-05167 GLC-05184 GLC-01021 GLC-01029 GLC-09361 GLC-05186 GLC-05182 GLC-08763 GLC-04924 GLC-09036 GLC-00978 GLA-07794 GLC-07865 GLA-00798 GLC-01042 GLC-01003 GLC-01008 GLC-05080 GLC-09161 GLA-07796 GLC-07467 GLC-05165 GLC-00993 GLA-00797 GLC-00984 242636 GLC-04883 GLC-01020 GLC-00980 GLC-13072 GLC-04880 GLC-00988 GLC-01027 GLA-00787 GLA-08774 GLC-01048 GLC-01010 GLC-00983 GLC-01007 GLC-01011 GLC-01015
16087-056CS 501.2 USD InStock
16087-056 89176-202 66009-553 16087-052 66009-551 66009-550 11311-216 16054-222 11311-218 82020-808 11311-214 76487-352 10798-854 16054-224 66009-546 76487-356 76487-354 76487-238 76487-236 66009-548 16087-058 16218-124 16087-044 76487-218 16087-042 16218-126 16218-128 10798-852 10798-850 89176-854 16054-214 76487-220 16054-212 76487-222 16087-048 66009-575 35100-129 93001-566 66009-567 66009-566 66009-565 66009-560 66009-563 66009-562 76209-748 66009-561 TNGLC0101 76487-242 76487-240 66009-557 66009-556 66009-555 89210-120 66009-554 76487-244 66018-574 66009-559 66009-558 TNGLC-05028 66175-515 11311-650 16087-090 TNGLC-05027 89210-118 75832-344 11311-648 16087-122 16087-088 36319-420 66175-520 18087-086 16087-128 16087-126 82020-162 16087-078 16218-134 16087-076 16218-136 16087-074 16087-072 93001-640 16087-070 11311-514 16218-130 16218-132 16054-208 76170-370 76170-374 76487-216 76539-342 66018-665 76539-344 76403-540 16087-068 76539-360 16087-066 76539-362 16087-064 16087-062 16087-060 16151-995 BAGLC07467ASY 82020-462
Sample Vials, Clear Borosilicate Glass, Screw-Thread
Vials
Clear, flint screw-thread sample vials offer maximum visibility.
  • Available with a variety of caps, or as vials only, to meet specific requirements
  • Pre-cleaned options save time and money
These general-purpose glass vials are made of Type I Borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty