Would you like to log in again?
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Supplier: Qorpak
GLA-00799 GLC-01058 GLC-07015 GLC-12579 GLC-01057 GLC-01063 GLC-05180 GLC-05646 GLC-01075 GLC-01053 GLC-06030 GLC-07291 GLC-08767 GLC-06789 GLC-08768 GLC-04885 GLA-07799 GLA-00801 242635 GLC-04886 GLC-01065 GLA-00802 GLC-07869 GLC-06161 GLC-04892 GLC-07294 242632 GLA-07798 GLC-01071 GLC-01059 GLC-05181 GLC-06788 GLC-06786 GLC-04917 GLC-06001 GLC-04918 GLC-01069 GLC-06029 GLC-01061 GLC-01076 GLC-01056 GLC-06031 GLA-05558 GLC-06787
35100-100CS 306.26 USD InStock
35100-100 11311-570 66175-518 35100-104 93000-912 35100-102 35100-106 89204-962 93001-538 36319-408 76170-372 76487-230 76487-234 76487-232 76076-732 76487-358 16087-100 93000-922 16087-084 89422-978 16054-210 93001-596 16087-082 89237-402 66175-540 16087-080 76170-368 11311-222 93000-906 11311-522 82027-246 93000-928 11311-224 10798-848 16054-216 10798-846 93001-766 76487-224 76487-246 76487-228 16087-104 76487-248 76487-226 16087-102
Sample Vials, Amber Borosilicate Glass, Screw-Thread, Qorpak®
Vials
Amber screw thread sample vials offer protection from harmful light and UV rays for light sensitive products.
These general purpose glass vials are made of Type I borosilicate glass, providing exceptional resistance to heat shock and chemical leaching.
Ordering information: Vials 36319-407 and 66175-540 are supplied pre-cleaned for volatiles. Vial 11311-522 is supplied ultra cleaned. Vials 93000-912, 93000-922, 93000-906, and 93000-928 are supplied vacuumed and ionized.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty