Would you like to log in again?
Qorpak® Black Phenolic Screw Caps, Pulp/Aluminum Foil Liner
Supplier: Qorpak
CAP-00285 CAP-00307 CAP-00301 CAP-00124 CAP-00094 CAP-00278 CAP-00232 CAP-00316 CAP-00177 CAP-00246 CAP-00114 CAP-00250 CAP-00162 CAP-00110 CAP-00290 CAP-00312 CAP-00090 CAP-00218 CAP-00213 CAP-00305 CAP-00270 CAP-00266 CAP-00259
11311-296PK 77.18 USD InStock
11311-296 16087-222 16087-232 16087-220 11311-298 11311-441 16087-230 11311-292 11311-304 11311-306 11311-288 11311-302 89204-930 76169-366 16087-218 16087-228 16087-238 16087-226 16087-214 16087-236 16087-224 16087-234 16087-212
Qorpak® Black Phenolic Screw Caps, Pulp/Aluminum Foil Liner
Closures Bottle Closures
Pulp/Aluminum Foil lined caps are typically used for food applications as they will not cause odor or taste problems
They can be used for non-acid, non-alkaline products. Great for organic solvents, chrome cleaners, brake fluids and mineral oils.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty