Would you like to log in again?
Black Polypropylene Screw Caps, Unlined, Qorpak®
Supplier: Qorpak
CAP-00367 CAP-00390 CAP-00394 CAP-00346 CAP-00413 CAP-00370 CAP-00401 CAP-00384 CAP-00362 CAP-00383 CAP-00396 CAP-00388 CAP-00325 CAP-00357 CAP-00333 CAP-00406 CAP-00404 CAP-00334 CAP-00350 CAP-00323 CAP-00381 CAP-00410 CAP-00399
16087-266CS 131.16 USD InStock
16087-266 16087-254 75796-210 16087-264 16087-275 16087-242 16087-262 16087-260 30617-122 11311-578 93001-598 93001-610 11311-574 11311-576 93001-604 93001-616 93001-600 93001-612 93001-602 82020-060 16087-268 16087-246 16087-256
Black Polypropylene Screw Caps, Unlined, Qorpak®
Closures Bottle Closures
Unlined polypropylene closures are autoclavable and are suitable for many dry products.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty