Would you like to log in again?
Cylinder Bottles, PVC, Narrow Mouth, Qorpak®
Supplier: Qorpak
PLA-05757 PLC-05774 PLC-05773 PLA-05758 PLC-05772 PLC-05772 PLC-05771 PLA-05759 PLA-05756
76470-978CS 449.58 USD InStock
76470-978 BAPLC05772CAR 30620-989 75877-328 16087-417 82023-684 76470-980 16087-414 76470-982
Cylinder Bottles, PVC, Narrow Mouth, Qorpak®
Bottles Plastic Bottles
Clear PVC Cylinder Bottles provide an excellent resistance to oxygen permeation, oils and good water barrier characteristics.
  • Transparent, rigid cylinders allow for easy visual identification of the amount of product inside
Narrow mouth cylinders are perfect for handling liquid samples in the lab. The shoulder style allows you to pour small quantities without spilling.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty