Would you like to log in again?
Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2221-0020 2221-0010
16093-022CS 755.52 USD InStock
16093-022 16093-011
Nalgene® Dispensing and Storage Jugs, Polypropylene, Thermo Scientific
Bottles Carboys
Autoclavable, leakproof jugs offer excellent chemical resistance.
  • Leakproof
  • Autoclavable
Note: Before autoclaving, set cap on top of jug without engaging the threads.
A large carrying handle allows for the use of rubber gloves for safe handling of corrosive liquids, as well as an offset pour spout for easy, accurate pouring.
Ordering information: Jugs are supplied with polypropylene screw closures. For replacement caps, see 16219-122 series
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty