Would you like to log in again?
Nalgene® Safety Dispensing Jugs, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2340-0050 2340-0020
16097-023CS 1355.52 USD InStock
16097-023 16097-012
Nalgene® Safety Dispensing Jugs, Low-Density Polyethylene, Thermo Scientific
Bottles Carboys
For the safe dispensing of liquids.
  • Leakproof
  • Polypropylene screw closures
For replacement caps, see cap 16225-234 in 16219-122 series.
Wide shoulder handle for easy carrying and bottom hand grip for additional support. Pour spout reduces splash. Spout plug is permanently attached to jug. Cap size: 83B.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty