Would you like to log in again?
Nalgene® Carboys with Handles, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2250-0020 2250-0050 2250-0130
16101-109CS 830.4 USD InStock
16101-109 16101-142 16101-084
Nalgene® Carboys with Handles, Polypropylene, Thermo Scientific
Bottles Carboys
Excellent for storing culture media, distilled water, and other solutions.
  • Leakproof
  • Autoclavable
For replacement caps, see cap 16225-234 in 16219-122 series.
Note: Autoclave using properly vented closures.
Graduated to contain in 1 gal. and 5 L increments. Carboys are equipped with two wide shoulder handles. Cap size: 83 mm. Accepts No. 13¹/₂ rubber stoppers.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty