Would you like to log in again?
NALGENE® Screw Caps, High-Density Polyethylene, Sterile, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
342151-0200 342151-0384 342151-0240
16101-098CS 115.2 USD InStock
16101-098 16057-316 16057-318
NALGENE® Screw Caps, High-Density Polyethylene, Sterile, Thermo Scientific
Closures Bottle Closures
For radiation-sterilization applications
Use with NALGENE® bottles. Not autoclavable.
Ordering information: Bulk packaged.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty