Would you like to log in again?
Nalgene® Heavy-Duty Rectangular Carboy, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2214-0050
16115-335CS 999.88 USD InStock
16115-335
Nalgene® Heavy-Duty Rectangular Carboy, Thermo Scientific
Bottles Carboys
The Nalgene® HDPE heavy-duty rectangular carboy is a space-saving option for storing and transporting reagents and other liquids.
The carboy is graduated to contain in 5 liter and 1 gallon increments.
The rugged carboy features large handles to enable easy use, even with gloved hands.
Delivery information: Supplied with valve-sealing, 70 mm closure.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty