Would you like to log in again?
Nalgene® Carboys with Handles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2210-0130 2210-0020 2210-0040 2210-0065 2210-0050
16117-062EA 344.58 USD InStock
16117-062 16117-040 16117-007 16117-018 16117-029
Nalgene® Carboys with Handles, Low-Density Polyethylene, Thermo Scientific
Bottles Carboys
Useful for storing and containment of chemicals and standards.
  • Wide shoulder handles for easy carrying and pouring
  • Supplied with ca
  • Cap size: 83B
  • Accepts No. 13¹/₂ rubber stopper
  • Leakproof
Ordering information: For replacement caps, see cap 16225-234 in 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty