Would you like to log in again?
Nalgene® Rectangular Carboys with Spigot and Handle, Fluorinated High-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
DS2327-0050 DS2327-0020
16117-985CS 291.21 USD InStock
16117-985 16117-984
Nalgene® Rectangular Carboys with Spigot and Handle, Fluorinated High-Density Polyethylene, Thermo Scientific
Bottles Carboys
Ideal for storing and dispensing a variety of solvents, and more aggressive reagents.
  • Carboys have molded-in graduations and stainless steel handle
For replacement caps, see cap 16219-106 in 16219-030 series.
Tough, rigid translucent carboy features fluorinated inside and outside surfaces to enhance long-term container performance by reducing solvent absorption and permeation while increasing chemical resistance.
Ordering information: Supplied with Tefzel® spigot and 100-415 fluorinated polypropylene screw cap.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty