Would you like to log in again?
Nalgene® Square Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2018-0500 2018-0125 2018-0030 2018-0060 2018-1000 2018-0250
16120-724CS 281.74 USD InStock
16120-724 16120-701 16120-708 16120-719 16120-728 16120-715
Nalgene® Square Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Designed for long-term storage of chemicals and specimens where sterility is not required.
  • Leakproof
  • Not autoclavable
Bottles provide excellent chemical resistance, and withstand temperatures from –100 to 120 °C (–148 to 248 °F). The square design is easy to handle, and saves space on benchtop or shelf, in refrigerators and freezers, and under the lab hood. Permanent graduations on all sizes.
Ordering information: Supplied with polypropylene screw caps assembled on bottles. For replacement caps for 16120-715 through -728, see 16219-122 series.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty